Would you like to log in again?
Heat Sealing Films, Clear
Supplier: Azenta Life Sciences
4TI-0540/80 4TI-0540 4TI-0541 4TI-0542 4TI-0542/S 4TI-0540/S
76637-788EA 1217.37 USD InStock
76637-788 76637-930 76637-828 76637-936 76637-836 76637-912
Heat Sealing Films, Clear
Seals Microplate Seals
The clear heat seal is an optically clear laminate film forms a peelable seal to polypropylene, polyethylene, polystyrene, polycarbonate and cyclic olefin copolymer (COC) SBS format plates.
  • Samples can be accessed by pre-piercing with a blade, needle or the pierce plate (4ti-0398)
  • Moderate solvent resistance and can be utilized for short term compound storage at room temperature
  • Temperature range: –80 to +80 °C
  • Non sterile
Optical clarity enables its use for sealing plates required for imaging use, including fluorescent detection methods such as qPCR and colorimetric assays.
Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. These clear heat seals form a complete seal to a plate enabling both low temperature uses, including low temperature storage, and high temperature uses, such as PCR (when used with a pressurized heated lid).
The sheet seals fit all standard SBS footprint plates from 12 to 1536 wells and are inter-leaved with paper sheets to help denote which side is the sealing side and aid removal of one sheet at a time from the pack.
Ordering information: Available as sheets for use with manual and semiautomated sealers such as the semi-automated sheet heat sealer. Also available in multiple roll formats compatible with specified automated heat sealers such as the automated roll heat sealer. Gamma treatment available upon request.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty