Would you like to log in again?
PCR Tube Strips, 8-Well, Azenta Life Sciences
Supplier: Azenta Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4TI-0753 4TI-0754/C 4TI-0754 4TI-0753/C
76637-996CS 154.04 USD InStock
76637-996 76638-128 76638-070 76638-062
PCR Tube Strips, 8-Well, Azenta Life Sciences
Tubes PCR Tubes
Strips of 8 polypropylene PCR tubes. Can be used alone or in conjunction with any 96-well PCR plate frame.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Low profile: <0.1 ml (100 µl) working volume, 0.2 ml (200 µl) total well capacity when used with sealing options
  • Can be used as individual strips, or in a 96 Well PCR Plate frame for easier handling
  • Compatible with standard multichannel pipettes
These low profile PCR tube strips are molded from virgin polypropylene under cleanroom conditions. They are available in either clear polypropylene for standard PCR techniques, or in white polypropylene for use in fluorescent detection applications like qPCR, as they give the highest sensitivity and consistency as all the fluorescence signal is reflected back to the detector.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty