Would you like to log in again?
Storage Microplates, 96 Round Well, Deep Well, Azenta Life Sciences
Supplier: Azenta Life Sciences
4TI-0120 4TI-0130
76638-136CS 300.72 USD InStock
76638-136 76638-010
Storage Microplates, 96 Round Well, Deep Well, Azenta Life Sciences
Microplates Deep Well Microplates
These 96 deep well storage microplates are suitable for many manual and automated protocols, such as the Illumina® library and sample preparations, due to the plates’ SBS footprint. Clear plate, perfect for storage applications.
  • Made from polypropylene, a material with very low biomolecular binding characteristics, a high temperature tolerance and resistance to many standard laboratory chemicals, including DMSO
  • Autoclavable and stackable
  • Free from RNase, DNase, human genomic DNA and endotoxins
  • Alphanumeric grid reference to aid well and sample identification
  • SBS footprint
  • Round wells, suitable for most applications as they reduce droplet effects and wicking
  • Temperature range for use: –80 to 120 °C
  • Suitable for adhesive and heat sealing, and as a collection plate from filter systems
U-shaped bottom plates are best for washing, mixing and pelleting and give high surface area.
Accessories information: 96 round well sealing cap mat, white silicone, for use with 4TI-0120 only. 96 round well sealing cap mat, clear silicone, for use with round 96 well microplates and deep well storage microplates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty