Would you like to log in again?
PCR Plates, 96-Well, Non-Skirted, Azenta Life Sciences
Supplier: Azenta Life Sciences
4TI-0750/W 4TI-0750/R 4TI-0750 4TI-0750/P 4TI-0750/G 4TI-0750/Y 4TI-0750-25 4TI-0750/B
76637-966CS 231.93 USD InStock
76637-966 76637-974 76638-222 76638-034 76638-048 76638-086 76638-084 76638-072
PCR Plates, 96-Well, Non-Skirted, Azenta Life Sciences
Microplates PCR Plates
Universal 96-well non-skirted plate designed for use on standard thermal cyclers. 0.2 ml wells, polypropylene, cut corner H12; working volume: <200 µl, total well capacity: 300 µl.
  • Subject to a strict QC procedure which includes a visual check and leak testing of every tube
  • Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Low profile 0.2 ml (200 µl) working volume, 0.3 ml (300 µl) total well capacity when used with sealing options
  • Maximum thermal conductivity for efficient heat transfer and precise thermal cycling
  • Raised well rims prevent cross contamination and facilitate effective sealing for reduced evaporation
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
  • Compatible with blocks designed for skirted PCR plates and standard multichannel pipettes
All of these PCR plates are molded from virgin polypropylene under ISO certified cleanroom conditions in a UK-based production facility and as such comply with the same stringent requirements as the FrameStar® range.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty