Would you like to log in again?
VersaHOCI™ Stabilized 0.065% Hypochlorous Acid, Berkshire
Supplier: Berkshire
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VersaHOCI™
VHOCL6Q
76638-264CS 99.78 USD InStock
76638-264
VersaHOCI™ Stabilized 0.065% Hypochlorous Acid, Berkshire
Cleaning Agents Disinfectants
VersaHOCl™ is an EPA registered, RTU (ready to use) one-step cleaner, disinfectant, and deodorizer. Made in the USA with no preservatives or fragrances, Berkshire’s VersaHOCl™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs.
  • Kills common microbiological contaminations on surfaces
  • Very low residue, saving time on residue removal
  • Wide range of material compatibility
  • No special disposal is required
  • Bottles individually bagged
  • Facilitates transfer disinfection into cleanroom
  • Can be used safely on all hard, non-porous surfaces
  • Ready-to-use; no mixing required
  • Not classed as corrosive
VersaHOCl™ is an EPA registered, RTU (ready to use) one-step cleaner, disinfectant, and deodorizer. Made in the USA with no preservatives or fragrances, Berkshire’s VersaHOCl™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs. Non-toxic with low odor and low residue, VersaHOCl™ establishes the safest standards in disinfectant technology. With an active ingredient of 0.065% hypochlorous acid, it’s non-corrosive and effective on hard, non-porous surfaces. This versatile disinfectant is ideal for use in controlled environments such as labs, cleanrooms, compounding pharmacies, and manufacturing facilities. It can also be used in areas such as hospitals, clinics, healthcare facilities, schools, and government and office buildings.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty