Would you like to log in again?
SatPax® Stencil Cleaner 100% Polyester Nonwoven Cleanroom Wipes, Berkshire
Supplier: Berkshire
SatPax®
SPXPNWZ00116 SPXPNWZ00116R
76638-274CS 467 USD InStock
76638-274 76638-270
SatPax® Stencil Cleaner 100% Polyester Nonwoven Cleanroom Wipes, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Berkshire SatPax® Stencil Cleaner Wipes with Zestron Vigon® SC 200 is a powerful combination of low lint wiping material and effective stencil chemistry unmatched in the industry. SatPax® Stencil Cleaner Wipes are safe, fast-drying, and odor-free.
  • Exceptional cleanliness
  • Smooth and durable with good wet strength
  • Non-shedding and low linting
  • Knife cut, perforated roll
  • Chemically compatible with common cleaning and disinfecting solutions
  • Increases cleaning efficiency and cleaning protocol consistency
  • No visible staining to stencil
Berkshire SatPax® Stencil Cleaner Wipes with Zestron Vigon® SC 200 quickly and effectively remove all types of solder pastes and adhesives used in electronic assembly manufacturing. These wipes are an ideal replacement for alcohol-based cleaners, which may not yield satisfactory cleaning results and are prohibited due to safety and environmental restrictions on VOC emissions or waste disposal issues.
Certifications: ISO Class 5 – 8. Cleanroom Class 100 – 100,000. RoHS I, II, and III, WEEE, and REACH SVHC.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty