Would you like to log in again?
Alsto DP2000 Disc Pulveriser
Supplier: Klen International
Alsto
402237 402234
76638-284EA 43389.25 USD InStock
76638-284 76638-282
Alsto DP2000 Disc Pulveriser
Mills
Simple, robust machine designed for fine grinding of a large variety of sample and ore types. One fixed and one rotating 200 mm grinding plate turning on a horizontal axis, with the sample drawer located at the bottom of the chamber.
  • A simple screw adjustment to control product sizing via plate gap setting
  • Powerful 4kW, 3 phase drive motor
  • Supplied as standard on a base plate suitable for bench/workstation mounting
  • Durable chilled cast iron disc plates for long life
The Alsto DP2000 Disc Pulveriser is a simple, robust machine designed for fine grinding of a large variety of sample and ore types. The unit comprises one fixed and one rotating 200 mm grinding plate turning on a horizontal axis. Internal surfaces of the grinding chamber are smooth and easily cleaned, with the sample drawer located at the bottom of the chamber.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty