Would you like to log in again?
Alsto JC5030 Jaw Crusher
Supplier: Klen International
Alsto
402249 402250
76638-298EA 300258.72 USD InStock
76638-298 76638-300
Alsto JC5030 Jaw Crusher
Mills
The Alsto JC5030 is the largest jaw crusher. A high capacity unit suitable for laboratory use and plant applications. Rugged construction and high reduction capabilities together with low maintenance requirements make it outstanding for a large variety of applications.
  • Large input particle size capability
  • 500×300 mm jaw opening allows for lump sizes of up to 250 mm
  • 30 mm product size allows for fast, efficient reduction with a high throughput of 15,000 kg/hr
  • Simple and effective single toggle design reduces maintenance and repair time
  • Fast and easy jaw gap adjustment, ensuring positive control of product sizing
  • Hard wearing jaw plates and side liners
  • Installation flexibility, allowing for many different applications
  • Full and effective drive guarding ensures operator safety
  • Reversible and interchangeable jaw plates for longer life
  • Hard wearing, easily replaceable jaw plates and side liners
The Alsto JC5030 jaw crusher is the largest crusher and is a high capacity unit suitable for laboratory use where very large particle sizes are encountered and also plant applications. It’s rugged construction and high reduction capabilities together with low maintenance requirements make it an outstanding workhorse for a large variety of applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty