Would you like to log in again?
Individual Access Heat Sealing, Azenta Life Sciences
Supplier: Azenta Life Sciences
59-2000
76638-864EA 7318.28 USD InStock
76638-864
Individual Access Heat Sealing, Azenta Life Sciences
Heat Sealers Microplate Heat Sealers
The 96 well Individual Access plate can be sealed in one step using Individual Access seals.
  • These seals result in individually sealed tubes that can be pierceable, allowing for sample access
Individual Access sealing procedure using the semi-automated sheet heat sealer (59-2000):
1. Place adapter in open drawer of sealer.
2. Place Individual Access plate on adapter.
3. Place Individual Access seal onto the plate.
4. Plate is automatically sealed, remove plate.
5. Remove the backing liner from the seal.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty