Would you like to log in again?
Automated Plate Seal Removers, Azenta Life Sciences
Supplier: Azenta Life Sciences
XPeel®
X-TAPE_2000 XP-A
76638-884EA 46042.32 USD InStock
76638-884 76638-890
Automated Plate Seal Removers, Azenta Life Sciences
Heat Sealers Microplate Heat Sealers
Our Automated Plate Seal Remover automatically removes seals from a wide range of microplate types with the single touch of a button, eliminating the need for repetitive, manual removal of plate seals and enables the adoption of the gold-standard operating model.
  • Compatible with a variety of full-plate seals, including heat and pressure applied seals
  • Uses proprietary Automated Plate Seal Remover Tape to de-seal microplates, up to 400 seal removals per roll
  • Eliminates cross contamination common with manual seal removal techniques
  • Supports Quality Control measures requiring samples to be sealed until their moment of use
  • When used as part of an integrated workflow, seal removal verification feature reduces plate handling errors
  • Can be used as a standalone system, or integrated into automated and robotic workflows via Serial RS232 remote interface
  • Capacity to remove up to 200 plate seals per hour
Can be used with a wide variety of microplates, including full-skirted PCR plates, low-base microplates and deep-well (0 to 2 ml) plates.
Certifications: CE marked.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty