Would you like to log in again?
Environmental Express® Oakton® pH 100 Portable pH Meters
Supplier: Antylia Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Environmental Express®
35613-25 35613-28 35613-27
76639-022EA 291.95 USD InStock
76639-022 76639-018 76639-020
Environmental Express® Oakton® pH 100 Portable pH Meters
Electrochemical Meters pH and Multi-Parameter Meters
Convenient, compact design with large display.
  • Measured parameter and temperature shown on display at same time
  • Utilizes same electrodes as Oakton 6+ series
  • Stability, calibration, and low-battery indicators
  • Waterproof and dust-resistant IP 57 rating – great for use under harsh environments
The new addition to the Environmental Express Oakton product line features updates to measuring water quality easier and faster. The larger display is 2.75×2.0" for better viewing angle, and has indicators for battery life, stability, and calibration. It also shows both the parameter and temperature measurements at the same time. The pH 100 meter measures pH, mV, ORP, and temperature.
The built-in microprocessor chip enables advanced functions, such as auto calibration, auto temperature compensation, parameter setting, self-diagnosis, auto power-off, and low-battery reminders. The meter uses advanced digital processing technology, intelligently improving the response time and accuracy of your measurements. Stable reading and auto lock display modes are available.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty