Would you like to log in again?
VWR® Heavy-Duty Absorbent Wipers
Supplier: Avantor
VWR®
AVTR1 AVTR3 AVTR2
76639-024CS 155.03 USD InStock
76639-024 76639-026 76639-030
VWR® Heavy-Duty Absorbent Wipers
Wipes Disposable Wipes and Towels Wipes for General Applications
VWR® Industrial absorbent wipers are used in many industries and are made from recycled materials. These wipers are designed to be durable and resistant to abrasion.
  • Durable and absorbent heavy-duty wipers
  • Lint-free
  • Strong either wet or dry
  • Flat, bulk in box
These top‑tier wipers, renowned for their exceptional cleanliness and absorbency, are designed to excel in demanding environments. Crafted with a high‑performance fiber blend, they boast unparalleled lint control, ensuring a flawless finish with minimal risk of contamination. Engineered to resist snagging on various surfaces, these wipes offer both durability and reliability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty