Would you like to log in again?
SpectraMax® Mini Microplate Readers, Multi-Mode, Molecular Devices
Supplier: Molecular Devices
SpectraMax®
5089097 5089098 5089098 SMAX-MINI-AFL SMAX-MINI-AL SMAX-MINI-AF 5089099 5089104 SMAX-MINI-AL 5089096 5089099 5089103 SMAX-MINI-AFL 5089334 5089097 5089334 SMAX-MINI-AF 5089096 5089102 5089102 5089100 5089104 5089100 5089103
76640-498EA 2460.72 USD InStock
76640-498 CA76640-496 76640-486 76640-500 CA76640-498 76640-488 CA76640-486 76640-502 CA76640-500 CA76640-488 CA76640-502 76640-504 76640-490 CA76640-490 76640-492 CA76640-492 76640-494 76640-496 CA76640-494 CA76640-504 76640-506 CA76640-506 76640-508 CA76640-508
SpectraMax® Mini Microplate Readers, Multi-Mode, Molecular Devices
Microplate Readers
The SpectraMax® Mini multi-mode microplate reader offers a budget friendly solution that gives you exactly what you need when you need it. With three modes of detection for UV/Vis absorbance, fluorescence and luminescence, the SpectraMax® Mini reader is user-upgradeable - from two to three modes, providing researchers with a versatile instrument that does not compromise on sensitivity or performance.
  • Industry leading software - enjoy an extensive library of protocols, advanced reader control, and easy data management and analysis with our 'best in class' SoftMax Pro software
  • Approachable design - effortless functionality provides users with everything they need to run their most crucial applications quickly and without complications
  • AutoPMT technology - allows a wide range of concentrations to be prepared and read on one plate, avoiding saturation (or the loss of signal) and saving you time and money
  • Expanded wavelength range - Xenon flash lamps allow you to excite samples in the UV range where halogen does not
  • Affordable - gain access to over 30 years of experience and technology in a compact, multi-mode reader that was built for researchers with sensitive budgets
  • Flexible - perform your favourite applications with a user-friendly reader that accommodates plate types from six to 384-well formats; up to three modes of detection measuring absorbance with NFC-tagged filters for fluorescence, luminescence, or both
  • Easy to use - save time on training, data generation, and data analysis, for faster results
Everything you need to run your most crucial applications quickly and easily.
Access your favourite applications, including ELISA, DNA and protein quantitation, cell viability, gene expression, and more with a compact, upgradeable microplate reader that does everything you need and lets you add detection modes in the future.
The SpectraMax® Mini reader provides tuneable absorbance, filter-based fluorescence and luminescence, temperature control, and easy analysis and data export with SoftMax Pro software.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty