Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ashing Furnaces with Fold Down or Lift Up Door, Models LV and LVT, Nabertherm

Supplier: Nabertherm

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nabertherm
LV095H1ON1-240 LV095K1ON1-240 LV055H1LN1-208 LV055K1LN1-240 LV055K1ON1-240 LV034H1ON1-120 LV055H1ON1-240 LV095H1ON1-208 LV055K1ON1-208 LV155K1LN1-208 LV095H1LN1-208 LV034H1ON1-120 LV035K1ON1-208 LV035H1ON1-208 LV034K1ON1-120 LV095K1LN1-208 LV155H1ON1-208 LV034K1ON1-120 LV095K1LN1-240 LV034H1LN1-120 LV155K1ON1-240 LV035H1LN1-240 LV155K1LN1-240 LV055K1LN1-208 LV034K1LN1-120 LV034H1LN1-120 LV035H1ON1-240 LV055H1ON1-208 LV034K1LN1-120 LV035K1LN1-240 LV095H1LN1-240 LV035K1LN1-208 LV095K1ON1-208 LV035K1ON1-240 LV055H1LN1-240 LV155H1ON1-240 LV035H1LN1-208 LV155H1LN1-240 LV155K1ON1-208 LV155H1LN1-208
76537-982EA 9058.85 USD InStock
76537-982 76537-984 76640-510 76640-512 76537-986 76537-988 76640-514 76538-008 76538-006 76538-028 76538-004 76538-026 76538-002 76538-024 76538-012 76538-034 76538-032 76538-010 76538-030 CA76640-514 CA76640-516 76537-990 76537-992 76537-994 76537-996 76537-998 CA76640-510 CA76640-512 76538-018 76538-016 76538-038 76538-014 76538-036 76538-000 76538-044 76538-022 76538-020 76538-042 76538-040 76640-516
Ashing Furnaces with Fold Down or Lift Up Door, Models LV and LVT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
The ashing furnaces LV 3/11 - LVT 15/11 are especially designed for ashing in the laboratory.

  • Maximum temperature: 1100 °C, heating from two sides
  • Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
  • Insulation made of non-classified fiber material, solid state relays provide for low noise operation
  • Housing made of sheets of textured stainless steel, dual shell housing for low external temperatures and stability
  • Optional fold down door (LV models) which can be used as work platform or lift up door (LVT models) with hot surface facing away from the operator
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive

A special air intake and exhaust system allows air exchange of more than 6 times per minute. Incoming air is preheated to ensure a good temperature uniformity.

Accessories information: Optional accessories include: Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; process control and documentation via VCD software package for monitoring, documentation, and control. Contact your Avantor representative for more information.

Ordering information: These ashing furnaces can be ordered in a variety of sizes and voltage options, with fold down door (L) or lift up door (LT). All models are rated up to 1100 °C maximum temperature. Control options include the B510 controller (5 programs with 4 segments each) or C550 contrller (10 programs with 20 segments each).

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty