Would you like to log in again?
Recombinant Oxoguanine Glycosylase
Supplier: New England Biolabs
M0464S M0464S
76640-796EA 78 USD InStock
76640-796 CA76640-796
Recombinant Oxoguanine Glycosylase
Enzymes
Bifunctional DNA glycosylase with DNA N-glycosylase and AP lyase activities
  • Thermostable
  • Isolated from a recombinant source
  • Supplied with 10X Reaction Buffer
Thermostable OGG is an archaeal 8-oxoguanine (8oxoG) DNA glycosylase which acts both as a N-glycosylase and an AP-lyase. The N-glycosylase activity releases the damaged purine (8-oxoguanine) from double stranded DNA, generating an apurinic (AP) site. The AP-lyase activity cleaves 3' to the AP site leaving a 5' phosphate and a 3'-phospho-alpha, beta-unsaturated aldehyde. Unlike some other DNA glycosylases, Thermostable OGG specifically recognizes and cleaves only 8oxoG and no other modified bases.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty