Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thermo Scientific™ Matrix™ Screw Top Tube Caps, Cap Trays and Cap Inserts

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Matrix®
4470BLK 4470WHI 4470RED 4477BLU 4477PIN 4470ORG 4470PUR 4477YEL 4477GRY 4477RED 4470YEL 4470BLU 4477WHI 4477GRE 4477PUR 4477 4470GRE 4470PIN 4470
76642-742CS 239.69 USD InStock
76642-742 76642-664 76642-920 76642-612 76642-842 76642-710 76642-776 76642-682 76642-872 76642-652 76642-640 76642-860 76642-806 76642-844 76642-888 76642-858 76642-924 76642-912 76642-758
Thermo Scientific™ Matrix™ Screw Top Tube Caps, Cap Trays and Cap Inserts
Closures Test/Sample Tube Closures
Simplify sample storage with these color-coded caps and inserts.

  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
  • ScrewTop caps are compatible with storage down to –196 °C

Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Cap options simplify sample identification:
Cap trays feature standard microplate format compatible with automated equipment
Caps are available in seven color options to enable sample identification
Color inserts snap into cap for a secure fit; inserts are removable
Inserts are available in six colors

All caps and inserts are supplied free from DNA, Rnase, Dnase, endotoxins and cytotoxins

Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015.
Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL)
The vials and closures were tested and met the requirements of USP <88> Class VI plastics.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty