Would you like to log in again?
Thermo Scientific™ Sample Storage Starter Packs
Supplier: Thermo Scientific
Matrix®
HHD-ST-WP1D-2 HHD-HSX-WP1D-2 HHD-HSX-WP1D-510 HHD-WP1D-2 HHD-HSX-WP1D-505
76894-076EA 12560 USD InStock
76894-076 76642-914 76642-702 76642-822 76642-644
Thermo Scientific™ Sample Storage Starter Packs
Vials
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs.
  • Eliminate the need to reformat non-2D cryotube samples
  • Secure sample tracking with 2D, 1D, and human readable codes
  • Scan 96 tubes in as little as 1 second with advanced and premium starter packs
  • Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance
Choose from five different starter pack options.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty