Would you like to log in again?
VWR® Plus Manual Defrost Freezers with Inner Doors
Supplier: Avantor
VWR®
VWR-HC-MFP-20P VWR-HC-MFP-17P VWR-HC-MFP-14P
76644-734EA 7454.54 USD InStock
76644-734 76644-738 76644-730
VWR® Plus Manual Defrost Freezers with Inner Doors
Freezers Laboratory / General Purpose Freezers
The inner doors on these Plus Series freezers protect stored substances with superior cooling retention.
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
These manual defrost laboratory freezers offer professional cold storage for scientific and industrial settings. The Plus Series includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: ETL Listed
Ordering information: All units come with a two-year parts and labor warranty and a five-year compressor parts warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty