Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Large Paraffin Wax Dispenser, Boekel Scientific

Supplier: Boekel Scientific
1456XL-2 1456XL
76644-920EA 4326 USD InStock
76644-920 76554-126
Large Paraffin Wax Dispenser, Boekel Scientific
Paraffin Wax Trimmers and Dispensers
The Boekel large wax dispenser is ideal for larger histology laboratories that require large volumes of paraffin wax when embedding tissue samples. The unit has a heated and filtered spigot to make sure the wax flows freely during operation and a stainless steel lid and chamber to stay corrosion free.

  • Industries largest wax capacity for high throughput use
  • Adaptive digital control system melts wax quickly and maintains temperature
  • Hot Start feature automatically turns on and starts melting wax
  • Bright easy-to-use digital display
  • 2-year warranty

This device can be used as the primary source of paraffin wax or to supplement tissue processors and embedding centers. The advanced digital control system is easy to use and has a timer based Hot Start system that ensures the wax is melted prior to operation. The low wax alarm in conjunction with its 28 L capacity and ability to quickly melt large volumes of wax ensures that you will have access to molten wax throughout the day. This wax dispenser will increase efficiencies for years, guaranteed with its two-year warranty.

Chamber dimensions (W×D×H) : 29.2×48.9×19 cm (11¹/₂×19¹/₄×7¹/₂")
Default temperature : 55 °C

Overtemp safety cutoff : 88 °C
Temperature stability : ±1.0 °C

Certifications: UL/CSA/CE – RoHS certified.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty