Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Portable Cell Counter

Supplier: Four E's USA
CCT100101
76644-930EA 8693.78 USD InStock
76644-930
Portable Cell Counter
Cell Counters and Flow Cytometers Electronic Cell Counters
Fast and compact cell counter. For measuring cell concentrations and viability.

  • Processes six samples with one slide
  • Max speed: 1×104 to 3×107 cells/ml
  • Cell diameter range: 5 to 200 µm
  • Cell viability detection range: 0 to 100%

Delivery information: Typically in stock, ready to ship.

Packaging: Shipped in sturdy cardboard box with foam packaging.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty