Would you like to log in again?
Freezing Point Osmometer
Supplier: Four E's USA
OSM100100
76644-946EA 36759 USD InStock
76644-946
Freezing Point Osmometer
Osmometers
The OSM100100 Osmometer measures the total osmolality of any aqueous solution by Freezing Point Depression (FPD).
  • Built-in touchscreen for intuitive operation
  • Small sample quantity required for testing
  • No proprietary consumables required
  • Refrigerant-free dual cooling system
  • Simple calibration procedure using 2 or more points
  • Built-in printer for test results
Designed for clinical and laboratory analysis of aqueous solutions, the osmometer is suitable to test urine, serum, or other bodily fluids. No proprietary consumables are required with a 0.5 ml sample tube used as the sample holder. The instrument utilizes a highly precise sensor for accurate measurement and high repeatability. Molar concentration, freezing point, and molar concentration ratio are all measured and recorded.
The multipoint calibration feature guarantees accurately calibrated results, allowing from 2 up to 30 total calibration points. The touchscreen enables intuitive user operation. A built-in printer provides convenient output of test results, with a variety of formatting options available. Data storage, access management, and password login for administrative functions ensure that authorized users can view and manage test result data securely.
Accuracy: ±2 mOsm/kg (≤400 mOsm/kg), ±1.0% (>400 mOsm/kg)
Accessories information: The instrument uses a 0.5 ml sample tube. Four E’s sample tube or Axygen PCR Tube PCR-05-C are compatible.
Delivery information: Supplied with an operating manual, inspection sheet, 1 pack of Four E's 0.5 ml sample tubes, rubber suction bulb, touch pen and a power cord.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty