Would you like to log in again?
Microscope Slide Storage Cabinets, Four E's USA
Supplier: Four E's USA
SSC100103 SSC200104 SSC200102 SSC200103 SSC100101 SSC200101 SSC200201 SSC200202 SSC200203 SSC100104 SSC200204 SSC100102
76644-964EA 173.29 USD InStock
76644-964 76644-952 76644-966 76644-954 76644-968 76644-956 76644-958 76644-960 76644-970 76644-962 76644-950 76644-972
Microscope Slide Storage Cabinets, Four E's USA
Slide Holders
Four E's is a leading manufacturer of microscope slide storage cabinets. Multiple colors and base styles available.
  • Store up to 5000 microscope slides per cabinet
  • Multiple colors available
  • Durable cold rolled steel construction with epoxy paint
  • 10 year warranty
These slide storage cabinets are designed for long term storage of large quantities of microscope slides in histology laboratories and hospitals. These cabinets have 6 drawers that hold up to 5000 standard 1×3" microscope slides. These cabinets come with interlocking system for safe stacking and can be stacked safely up to 12 units high. They are made of cold rolled steel with epoxy powder coating. The drawers are designed to slide in and out smoothly. Each drawer comes with two foam blocks to maintain slides upright when drawers are partially filled. Two different base styles are available, with or without legs. Four colors available.
Ordering information: Cabinets and bases are sold separately. Ten-year warranty.
Delivery information: Includes foam drawer inserts.
Packaging: Packaged in sturdy cardboard box.Packaged in sturdy cardboard box.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty