Would you like to log in again?
Oligo Pro II Reagents
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DN-400-0001 DN-475-1000 DN-475-1000 GP-435-0100 GP-435-0100 DN-465-1000 DN-400-0001 DN-415-0250 DN-465-1000 DN-415-0250
76645-024EA 240.8 USD InStock
76645-024 76645-352 76645-362 CA76645-060 76645-360 CA76645-360 76645-060 CA76645-362 CA76645-352 CA76645-024
Oligo Pro II Reagents
Electrophoresis Systems Capillary Electrophoresis Systems
ssDNA and ssRNA oligonucleotide purity and QC analysis.
  • High separation resolution - provides accurate assessments of oligonucleotide purity, allowing for n-1 resolution through 60-mers
  • Ready-to-use reagents - easy system setup and minimal time lost to instrument preparation, improves laboratory throughput and efficiency
  • Capillary gel electrophoresis - UV detection - No dye labeling needed
  • Allows for direct detection of both ssDNA and ssRNA
  • High throughput - load up to three 96-well plates for automated analysis
The Agilent Oligo Pro II system provides quality control of oligonucleotides used in downstream applications where knowledge of synthesized oligo purity is critical. The OLIGEL ssDNA gel and associated ssDNA OLIGEL Buffer delivers accurate oligo purity analysis of ssDNA and ssRNA with n-1 resolution through 60 nt in length.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty