Would you like to log in again?
Capillary Arrays
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M5310AA-002 M5310AA-001 M5311AA-003 M5310AA-002 M5312AA-001 A2300-9650-3355 A2300-1250-2247 A2300-4850-3355 A2300-9650-5580 M5312AA-001 M5312AA-002 M5320AA-001 M5311AA-002 M5320AA-002 M5310AA-003 M5320AA-002 A2300-1250-3355 M5320AA-001 A2300-4850-3355 M5311AA-002 A2300-9650-3355 M5311AA-003 M5311AA-001 M5310AA-001 M5310AA-003 A2300-9650-5580 A2300-1250-2247 M5311AA-001 M5312AA-002 A2300-1250-5580 A2300-1250-3355 A2300-1250-5580
76645-312EA 3953.85 USD InStock
CA76645-168 76645-312 76645-234 76645-178 76645-078 76645-198 CA76645-404 CA76645-208 76645-074 76645-092 CA76645-092 76645-208 CA76645-074 76645-404 CA76645-198 CA76645-078 CA76645-178 CA76645-234 CA76645-312 76645-168 76645-144 76645-386 76645-462 76645-440 76645-064 CA76645-418 76645-418 CA76645-064 CA76645-462 CA76645-440 CA76645-144 CA76645-386
Capillary Arrays
Electrophoresis Systems Capillary Electrophoresis Systems
Capillary arrays for fragment analyzer and ZAG systems.
  • Array cartridges are robust and have a long lifespan, keeping your instrument on-task from one project to the next
  • Short arrays provide robust separation for most samples
  • Speed up separation times and increase overall QC throughput with ultra-short arrays
  • Enhance separation resolution and focus nucleic acid analysis with long arrays
The fragment analyzer systems offer a selection of capillary arrays to optimize your QC workflow by separating nucleic acids via parallel capillary electrophoresis. Arrays vary in capillary length, providing distinct benefits to a wide range of workflows.
These arrays address separation speed and separation resolution. 'Short' capillary arrays are the most versatile as they are compatible with all consumable kits. Run times can be decreased with 'Ultra-Short' arrays, and 'Long' arrays enhance separation resolution. Some kits have been designed for a specific array for optimal performance.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty