Would you like to log in again?
Bioanalyzer Protein Analysis
Supplier: Agilent Technologies
5067-1517 5067-1518 5067-1518 5067-1517 5067-1516 5067-1516 5067-1515 5067-1515
76645-464EA 1538.32 USD InStock
76645-464 76645-232 76645-298 CA76645-192 76645-192 CA76645-232 CA76645-298 CA76645-464
Bioanalyzer Protein Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
Electrophoretic separation of proteins and peptides up to 230 kD.
  • Flexibility to choose the appropriate kit based on the sizing range
  • Minimal sample consumption is required, with only 4 µl of sample needed for an accurate analysis
  • Results are achieved rapidly, with the automated analysis of 10 samples performed in approximately 30 minutes
  • Complete data analysis may be performed in one assay, providing sizing, quantitation, and purity information
  • Provides sensitivity comparable to coomassie blue stains across a large linear dynamic range
  • Meet GMP and GLP requirements with security pack, an optional software add-on feature that enables 21 CFR Part 11 compliance
Protein electrophoresis using the 2100 bioanalyzer system is a fast, automated, objective, and flexible solution for protein and peptide characterization, quality control, and impurity detection. The agilent protein 80 and protein 230 assays provide sensitivity comparable to coomassie blue stains.
The system eliminates handling of SDS-PAGE slab gels, staining, or imaging steps for a more efficient workflow. Sample types evaluated using the bioanalyzer system include protein lysates, purified protein and peptides, reduced and non-reduced antibodies, and protein stability tests.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty