Would you like to log in again?
Mineral Oil Dropper Bottle
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FS-SMO15 FS-SMO15
76645-242EA 61.28 USD InStock
76645-242 CA76645-242
Mineral Oil Dropper Bottle
Lubricants
Used as an overlay on DNA samples to minimize evaporation.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty