Would you like to log in again?
Femto Pulse 12-Capillary Array
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A1600-1250-2240 M5330AA-001 M5330AA-001 A1600-1250-2240
76645-264EA 3255.1 USD InStock
76645-264 CA76645-190 76645-190 CA76645-264
Femto Pulse 12-Capillary Array
Electrophoresis Systems Capillary Electrophoresis Systems
12-Capillary Array for analysis on the Femto Pulse system.
  • High resolution - optimized length for enhanced resolution; Run Multiple Samples - Run up to 12 samples at same time
  • Low sensitivity - integral part of the Femto Pulse system aiding in low sensitivity detection
The Femto Pulse 12-Capillary Array is used with the Genomic DNA 165 kb kit, the Ultra Sensitivity NGS kit (1 to 6000 bp), the Ultra Sensitivity RNA kit, and the 55 kb BAC kit. It has a separation or effective length of 22 cm, with an overall length of 40 cm. The 12 parallel capillary array is ideal for lower throughput laboratories.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty