Would you like to log in again?
High Sensitivity RNA ScreenTape Analysis
Supplier: Agilent Technologies
5067-5580 5067-5581 5067-5580 5067-5579 5067-5579 5067-5581
76645-286EA 127.93 USD InStock
76645-286 76645-260 CA76645-182 CA76645-260 76645-182 CA76645-286
High Sensitivity RNA ScreenTape Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
Reliable, reproducible, and objective measurements of RNA samples of limited abundance.
  • Utilizes RINe – a user independent algorithm for accurate and objective assessment of eukaryotic or prokaryotic total RNA integrity
  • Fast results are obtained in approximately 2 minutes per sample
  • Minimal sample consumption is required as only 2 µl of sample is needed
  • Detection of RNA sample with limited abundance down to 100 pg/µl is possible
  • Objective quality control of precious RNA samples is enabled within RNAseq, microarray analysis, and quantitative RT-PCR workflows
  • Ready-to-use consumables provide excellent reproducibility
  • No cross contamination and sample carryover by using single lanes and individual loading tips.;Partially used ScreenTape devices can be utilized for completion at a later point in time
High Sensitivity RNA ScreenTape analysis provides efficient and reliable separation of total RNA samples of limited abundance down to 100 pg/µl. Compatible with eukaryotic and prokaryotic samples, the assay provides quality, quantity, and sizing information. Objective evaluation of RNA degradation is delivered with an RNA Integrity Number (RINe).
Results are obtained quickly, as fast as 2 minutes per sample. Accurate analysis is possible with only 2 µl of sample.
The assay is compatible with all Agilent TapeStation instruments.
Note: Some products within this assay contain SYBR™ Gold, which is licensed from Life Technologies Corporation for use in research and development only. SYBR™ is a registered trademark of Life Technologies Corporation.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty