Would you like to log in again?
Genomic DNA Kits
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DNF-468-0500 DNF-467-0500 DNF-270-0240 DNF-270-0240 DNF-488-0500 DNF-270-0500 DNF-270-0500 DNF-488-0500 DNF-468-0500 DNF-488-1000 DNF-488-1000 DNF-467-0500
76645-374EA 3628.13 USD InStock
CA76645-104 CA76645-106 76645-374 CA76645-228 76645-460 CA76645-290 76645-290 CA76645-460 76645-228 76645-106 CA76645-374 76645-104
Genomic DNA Kits
Electrophoresis Systems Capillary Electrophoresis Systems
Genomic DNA quality control of whole genome libraries, including sizing of gDNA extractions.
  • Broad concentration range - The HS gDNA 50 kb kit has a concentration range of 0.3 to 12 ng/µl, while the gDNA 50 kb kit covers 25 to 250 ng/µl
  • High and standard sensitivity kits - Minimize sample dilutions and choose the kit that has the right input concentration range for your needs
  • Reliable Sizing - Sizing range from 75 bp to 60 kb ensures accurate and precise sizing of gDNA samples
  • Low sample volume - Only 1 to 2 µl of sample is required, minimizing sample loss to QC steps
The genomic DNA kits are available for the automated assessment of gDNA sizing and integrity on the fragment analyzer systems. The kits are designed for quality control of gDNA extraction samples used in whole genome sequencing, metagenomics, and large structural variant analysis. They are also suited for the analysis of degraded DNA, such as formalin-fixed parafilm embedded (FFPE) samples.
Sample preparation is simplified by eliminating dilution steps with separate genomic DNA kits for high (HS) and low concentrated gDNA samples. A broad sizing range allows researchers to accurately size gDNA samples through 60 kb.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty