Would you like to log in again?
Qualitative DNA Kits
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DNF-810-0500 DNF-935-K0500 DNF-905-0500 DNF-930-0240 DNF-920-0240 DNF-930-0500 DNF-910-K1000 DNF-810-0125 DNF-810-0240 DNF-915-K1000 DNF-905-K0500 DNF-905-K0500 DNF-920-K0500 DNF-910-K0500 DNF-935-K1000 DNF-905-0240 DNF-810-0125 DNF-930-K0500 DNF-915-K0500 DNF-905-0500 DNF-920-0240 DNF-905-0240 DNF-930-K0500 DNF-915-K1000 DNF-930-0500 DNF-930-K1000 DNF-905-K1000 DNF-810-0240 DNF-905-K1000 DNF-910-K1000 DNF-935-K1000 DNF-935-K0500 DNF-930-0240 DNF-920-K0500 DNF-915-K0500 DNF-910-K0500 DNF-810-0500 DNF-930-K1000
76645-256EA 218.23 USD InStock
CA76645-388 CA76645-366 CA76645-202 76645-256 76645-278 76645-058 CA76645-148 CA76645-126 76645-310 CA76645-402 76645-330 76645-010 76645-252 76645-050 CA76645-050 CA76645-252 CA76645-010 CA76645-330 76645-402 76645-148 76645-126 CA76645-310 CA76645-256 CA76645-058 CA76645-278 76645-202 76645-366 76645-388 76645-222 76645-068 76645-342 CA76645-116 76645-044 CA76645-044 76645-116 CA76645-342 CA76645-068 CA76645-222
Qualitative DNA Kits
Electrophoresis Systems Capillary Electrophoresis Systems
DNA fragment analysis for PCR, genotyping, and microsatellite applications.
  • Broad concentration range - all kits have a concentration range of 0.5 to 50 ng/µl
  • Reliable sizing - six different kits to choose from with a sizing range varying from 35 to 20,000 bp
  • Low sample volume - only 2 µl of sample is required, minimizing precious sample loss to QC steps
  • Quick separations - electrophoresis times as short as 15 minutes
  • High resolution - separate fragments under 300 bp differing by only 3 bp
The qualitative kits provide automated and accurate sizing with relative quantification of DNA fragments. Common uses include the analysis of PCR amplicons, simple sequence repeats (SSRs)/microsatellites, and genotyping. There are six different kits to choose from based on the size of the DNA or PCR fragments, ranging from 35 to 20,000 bp. Specially formulated gels, markers, and ladders are designed for each kit enabling reliable sizing over a broad range with varying applications. High resolution kits enable the separation of fragments under 300 bp with a 3 bp difference.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty