Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5X Inlet Buffers

Supplier: Agilent Technologies
DNF-325-0075 DNF-325-0010 DNF-325-0010 DNF-325-0075
76645-354EA 29.03 USD InStock
76645-354 CA76645-390 76645-390 CA76645-354
5X Inlet Buffers
Buffers Electrophoresis Buffers Running Buffers
  • Assay type: Small RNA
  • Buffer method: Dilution
  • Volume: 1 µl

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty