Would you like to log in again?
TapeStation Systems Parts and Accessories
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5188-8047 5067-5599 5067-5598 5188-8047 5067-5158 G2991-40007 5067-5783 5067-5598 5067-5599 5067-5786 G2992-40046 5067-5158 5067-5786 G2992-40042 5067-5783
76645-420EA 56.98 USD InStock
CA76645-038 76645-420 CA76645-478 CA76645-314 76645-474 76645-066 76645-142 76645-170 CA76645-170 76645-448 CA76645-142 CA76645-066 76645-314 76645-038 76645-478
TapeStation Systems Parts and Accessories
Electrophoresis Systems Capillary Electrophoresis Systems
Accessories and spare parts for the 4150 and 4200 TapeStation systems like plates & foil seals, loading tips, TapeStation test tape, needle cartridge.
  • 8-well PCR tube strips or 96-well sample plates are available depending on sample throughput, bringing added flexibility
  • Needle cartridges enable fast and simple needle exchange to ensure proper maintenance of the TapeStation systems
  • TapeStation test tape is available for troubleshooting and running system diagnostics tests, the tapes are reusable and expire after two years
  • Loading tips are pre-packed and ready to use
Our portfolio of TapeStation systems spare parts and accessories features everything needed to keep your lab productive. Maximize the efficiency of your TapeStation systems today with our genuine agilent accessories.
The portfolio features additional needle cartridges and TapeStation test tape. In addition, if running short on spare parts, extra loading tips are also available, along with 96-well sample plates and foil seals. For enhanced flexibility we also offer 8-well PCR optical tube strips and caps.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty