Would you like to log in again?
Sol-M® Hypodermic Needles
Supplier: Sol-Millennium
Sol-M®
111915B 111810B 112658 112510B 110022FNSB 112510B 113010 112210 112506B 112310B 112010 112115 112506B 112612 113005B 112506 112715B 111915 112515B 112715 112015 112215 112015B 111810 113005 112015B 112510 112310B 111815 112515 112310 111810B 112115B 112715B 112705 111915B 111910 112115B 110022NSB 112215B 111615 112215B 112110 110022FNSB 113005B 110022NSB 112638 112358
76645-653CS 622.56 USD InStock
76645-653 76645-631 76645-652 76645-630 76582-380 76582-384 76582-362 76582-360 76582-382 76582-366 76582-388 76645-609 76645-608 76582-386 76582-364 76645-605 76645-627 76582-368 76645-626 76645-604 76645-648 76645-603 76645-602 76645-601 76645-645 76645-600 76645-643 76645-642 76645-641 76645-640 76582-370 76582-392 76582-390 76582-396 76582-374 76582-394 76582-372 76582-378 76582-376 77007-664 76645-637 76645-636 76582-358 76645-635 76645-634 76645-655 77392-158 76645-654
Sol-M® Hypodermic Needles
Syringe Needles
The Sol-M hypodermic needle product line features a variety of needle length and gauge sizes.
  • Triple-bevel lubricated stainless steel bevel that minimizes patient discomfort
  • Color-coded hubs for easy gauge identification
  • Compatible with all standard luer-lock and slip tip syringes
Sol-Millennium® offers a wide range of hypodermic needles with an emphasis on ease of use and patient comfort.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty