Would you like to log in again?
Sol-M® Quincke Spinal Needle
Supplier: Sol-Millennium
Sol-M®
111922500NSB 111922700NSB 111922150NSB 111922315NSB 111925200NSB 111918315NSB 111925315NSB
76645-620CS 4787.24 USD InStock
76645-620 76645-618 76645-616 76645-614 76645-612 76645-622 76645-610
Sol-M® Quincke Spinal Needle
Syringe Needles
The anesthesia needles are intended for injection of local anesthetics into the spinal and epidural spaces of a patient to provide regional anesthesia.
  • Bulk
  • Non sterile
The administration of the spinal anesthesia allows rapid anesthesia onset and the placement of an epidural catheter allows for bolus injections or continuous infusion of local anesthetics or other drugs into the epidural space.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty