Would you like to log in again?
Sol-M® Standard Syringes without Needle
Supplier: Sol-Millennium
Sol-M®
P180005ST P180003STNSB 180060CT P180011LDS P180011 P180010NSB P180005NSB P180001PPNSB P180003NSB P180003ST 180060CTNSB P180030NSB P180020NSB P180020ST P180060NSB P180060 P180010STNSB P180011LDSNSB P180020 P180010ST P180003 P180005 P180005STNSB P180010 P180060ST P180020STNSB P180001PP P180030
76582-818BX 42.7 USD InStock
76582-818 76645-674 76645-672 76645-670 76582-800 76582-820 76645-668 76645-666 76645-688 76582-802 76582-824 76645-686 76582-808 76645-684 76582-828 76582-806 77021-008 76645-682 76645-680 76582-792 76582-630 76582-796 76582-794 76582-814 76645-656 76645-678 76582-812 76645-676
Sol-M® Standard Syringes without Needle
Syringes Syringes, Medical
3-Piece syringes accurately measure liquid medications and are available in a range of sizes and connection combinations to address a wide variety of clinical needs.
  • High-transparency barrel to easily visualize contents
  • Silicone lubrication ensures smooth plunger movement
  • Plunger backstop that prevents accidental withdrawal of plunger from the barrel
  • Large finger grips allow for easy, controlled injections
Available in Luer lock, Luer slip tip and catheter tip.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty