Would you like to log in again?
Sol-Care® Safety Needles
Supplier: Sol-Millennium
Sol-Care®
SN2558B SN2215B SN2110 SN2515 SN2315 SN2558 SN2510B SN2210 SN3012 SN2310 SN2515B SN2712 SN1815B SN2115 SN2015 SN2510 SN2215 SN1815 SN2115B
76582-928BX 29.96 USD InStock
76582-928 76582-918 76582-926 76582-916 76582-908 76582-920 76582-932 76645-704 76582-910 76645-714 76645-724 76582-930 76582-924 76645-722 76582-914 76582-922 76645-710 76582-912 76645-720
Sol-Care® Safety Needles
Syringe Needles
The Sol-Care safety needle is designed to help prevent needlestick injuries. Every needle is equipped with a pivoting safety shield mechanism that locks in place to lower the risk of accidental injury.
  • Two options for protective arm activation: Hard surface activation and thumb activation
  • Simple one-handed safety shield mechanism; audible click indicates safety mechanism is secure and locked in place
  • Protective arm does not obstruct visualization of injection site
  • Bevel-up needle allows for low-angle injections
Hubs, protective shields and packaging are color-coded for immediate identification of needles gauges (ISO-6009 in a compliance with criteria given by OSHA).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty