Would you like to log in again?
Automated T-Maze
Supplier: Maze Engineers
1403 1404
76669-828EA 11500 USD InStock
76669-828 76669-826
Automated T-Maze
Animal Supplies Animal Environmental Enrichment
The automated version of the T-Maze explores memory and cognition using spontaneous alternation or rewarded alternation.
  • Quiet automated pnaeumatic doors minimize human intervention
  • Easy to clean acrylic construction
The automated T-Maze uses a return to start module and automated pneumatic downward facing doors to control the subjects route without human intervention. Automated reward delivery and video tracking integration maximize ease of use and automated experiments.
Ordering information: The Automated T-Maze requires software to function (see accessory options). Software packages are designed for use with the Automated T-Maze and video tracking software such as EthoVision or ANY-Maze.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty