Would you like to log in again?
Automated Radial Arm Maze
Supplier: Maze Engineers
1901 1902
76669-838EA 16500 USD InStock
76669-838 76669-836
Automated Radial Arm Maze
Animal Supplies Animal Environmental Enrichment
The automated 8 arm Radial Mazes creates an entirely automated environment with automated doors, reward delivery, and video tracking integration.
  • Quiet automated pnaeumatic doors minimize human intervention
  • Easy to clean acrylic
  • Matte, non-reflective surfaces
  • Odor-free
The automated 8 Arm Radial maze is an apparatus widely validated for spatial learning tasks using external cues and arm baiting. Automated doors, reward delivery, and video tracking integration minimize anxiety and fear created by the human experimentor while collecting high volumes of data.
Ordering information: The Automated Radial Arm Maze requires software to function (see accessory options). Software packages are designed for use with the Automated Radial Maze and video tracking software such as EthoVision or ANY-Maze.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty