Would you like to log in again?
Morris Water Mazes
Supplier: Maze Engineers
3103 3101
76669-846EA 2520 USD InStock
76669-846 76669-844
Morris Water Mazes
Animal Supplies Animal Cages
The Morris water maze (MWM) and additional accessories are used to assess spatial learning and memory in rodents.
  • Made of plastic, and is availble in white and blue
  • An adjustable platform is included with every MWM order
The maze consists of a large circular pool filled with water and a hidden platform. The subject is placed in the pool and is required to find the hidden platform in order to escape the water.
Accessories information: Optional steel frame with casters is designed to fit the MWM with side drain. The optional radial arm inserts, T-maze inserts, and Y-maze inserts are designed to fit the MWM with closed ends. The optional MWM heater will heat the MWM from 0 to 90 ℉ with 1 ℉ differential.
Ordering information: The Morris Water Maze is available in a white or blue plastic finish, with a side drain or bottom drain. When ordering, please specify the desired options. The MWM adjustable platform for mouse has a top width of 4 cm to 9 cm available in 1 cm increments. The MWM adjustable platform for rat has a top width of 10 cm to 16 cm available in 1 cm increments. When ordering, please specify the width required, as well as a round or square shape. Please enqiure for customization.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty