Would you like to log in again?
T-Maze
Supplier: Maze Engineers
3403 3402 3401
76669-920EA 1720 USD InStock
76669-920 76669-916 76669-918
T-Maze
Animal Supplies Animal Environmental Enrichment
Similar to the Y-Maze, T-Maze is a behavioral test used to assess spatial learning and memory in rodents.
  • Acrylic construction
  • Easy assembly and disssembly with a removable base
  • Easy to clean
The T-Maze is a behavioral test used to assess spatial learning and memory in rodents. The T-Maze consists of a starting arm and two goal arms, arranged in the shape of a 'T'. The goal arms can be baited with food or other rewards. The subject's movements are then tracked, and different measures such as the number of arm entries, the time spent in each arm, and the number of errors made can be used to assess the subject's memory and spatial navigation abilities.
The T-Maze for mouse measures 66 cm across the T-Maze. The T-Maze for rat measures 110 cm across the T-Maze. The T-Maze with central partition for mouse measures 70 cm across the T-Maze, with a central partition option.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty