Would you like to log in again?
Sociability Cages/Chambers
Supplier: Maze Engineers
3801 3802
76669-978EA 2653.34 USD InStock
76669-978 76669-980
Sociability Cages/Chambers
Animal Supplies Animal Cages
The 3-chamber, sociability cage is used to study rodent socialization.
  • Easy to clean acrylic walls and floors, floor cues
  • Stainless-steel grids or perforated stainless-steel to forge an aversive stimulus
  • Removable doors to establish biased and unbiased conditioned place preference testing
The 3-chamber sociability cage is designed to assess multiple variables that may be altered to change the premises of the experiment. The design permits socialization but disallows aggravated socialization, so that auspicious and accurate data may be collected.
Ordering information: When ordering, please specify acrylic rod or metal rod cage design.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty