Would you like to log in again?
Light/Dark Boxes
Supplier: Maze Engineers
4102 4101
76670-004EA 1853.34 USD InStock
76670-004 76670-002
Light/Dark Boxes
Animal Supplies Animal Environmental Enrichment
The Light/Dark box is extensively used to test anxiety-like behavior in rodents by assessing a subject’s reaction to the aversive and anxiety-prone context of being in a brightly illuminated area.
  • Acrylic construction
  • Easy to clean
  • Odor-free
  • Matte finish to remove shine
  • Easy assembly and disssembly with a removable base
Animals in anxiety models spend less time in the bright chamber, while in comparison, animals receiving anxiolytic treatment show an increase in the time spent in the bright area. The maze has been used to study anxiety and the effect of diseases and treatments on anxiety while different studies have adapted this apparatus in order to collect data regarding different aspects of anxiety-like behavior.
The Light/Dark box for mouse features a 25×40 cm light chamber, 17.5×40 cm dark chamber, and a 5 cm door. The Light/Dark box for rat features a 30×40 cm light chamber, a 20×40 cm dark chamber, and a 10 cm door.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty