Would you like to log in again?
Tail Suspension Tests
Supplier: Maze Engineers
4201 4202 4204
76670-014EA 2126.67 USD InStock
76670-014 76670-018 76670-016
Tail Suspension Tests
Animal Supplies Animal Environmental Enrichment
The tail suspension test is a standard test of depression in rodents.
  • Acrylic construction
  • Non reflective surface
  • Easy customization
Since its creation in 1985, the tail suspension test is a standard test of depression (Steru et al., 1985). The standard protocol is to hang mice to the apparatus by their tail.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty