Would you like to log in again?
Sucrose Preference Test Apparatus
Supplier: Maze Engineers
4432 4431
76670-048EA 3986.67 USD InStock
76670-048 76670-046
Sucrose Preference Test Apparatus
Animal Supplies Animal Environmental Enrichment
The Sucrose Preference Test (SPT) is a behavioral test used to assess anhedonia behaviors.
  • Acrylic construction
  • Non reflective surface
The Sucrose Preference Test (SPT) is a behavioral test used to assess anhedonia, a state characterized by the inability to experience pleasure, in small animals such as rodents. The test is based on the assumption that subjects will naturally prefer sweet solutions, such as sucrose, over plain water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty