Would you like to log in again?
Hole Board Tests
Supplier: Maze Engineers
4502 4501
76670-052EA 2653.34 USD InStock
76670-052 76670-050
Hole Board Tests
Animal Supplies Animal Environmental Enrichment
The modified hole board is a behavioral test used in neuroscience to assess multiple aspects of unconditioned behavior, cognition, and social interaction.
  • Acrylic construction
  • Non-reflective surface
  • Available with 4 or 16 hole removable inserts
The Maze Engineers hole board comes with inserts of either 4 or 16 holes. The inserts are easily removable and can be used as an open field chamber for secondary experiments. The entire wall apparatus can be removed for easy cleaning.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty