Would you like to log in again?
Hebb-Williams Mazes
Supplier: Maze Engineers
4701 4702
76670-088EA 3853.34 USD InStock
76670-088 76670-086
Hebb-Williams Mazes
Animal Supplies Animal Environmental Enrichment
The Hebb-Williams Maze (HWM) is a type of behavioral test used to assess spatial learning and memory in rodents.
  • Acrylic construction
  • Non reflective surfaces
The HWM consists of a central platform with several arms radiating out from it. The maze is made of acrylic and the arms can be baited with food or other rewards. The test is typically conducted by placing the animal on the central platform and allowing it to explore the maze. The animal’s movements are then tracked, and different measures such as the number of arm entries, the time spent in each arm, and the number of errors made can be used to assess the animal’s spatial learning and memory abilities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty