Would you like to log in again?
Puzzle Boxes
Supplier: Maze Engineers
4703 4704
76670-092EA 2393.34 USD InStock
76670-092 76670-090
Puzzle Boxes
Animal Supplies Animal Environmental Enrichment
The puzzle box was described by Deacon and colleagues in 2011 as a variant of the work of Galsworthy et al., (2002, 2005).
  • Acrylic construction
  • Non-reflective surfaces
Subjects are placed in a brightly lit compartment and quickly develop a preference for a smaller, dark goal compartment due to light/dark motivation. Subjects are challenged with various interruptions of increasing difficulties, and are tasked to adopt solutions to each new problem.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty