Would you like to log in again?
Lashley III Maze
Supplier: Maze Engineers
4705 4706
76670-096EA 2653.34 USD InStock
76670-096 76670-094
Lashley III Maze
Animal Supplies Animal Environmental Enrichment
The Lashley III maze is a test used for learning and memory. It utilizes a low stress environment and looks into route learning of a maze under repeated trials.
  • Acrylic construction
  • Non-reflective surfaces
In typical protocols, no stress, food deprivation, or other stimuli are used. A start box, a maze body, and a goal box are contained in the complete apparatus. The goal box can be filled with bedding similar to a home cage to reward the subjects, and to motivate them toward completion of the task. A large red base is not included in the order, but can be ordered upon request.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty