Would you like to log in again?
Bite Test Chambers
Supplier: Maze Engineers
4810 4809
76670-114EA 920 USD InStock
76670-114 76670-116
Bite Test Chambers
Animal Supplies Animal Environmental Enrichment
The Zebrafish bite test evaluates cerebral lateralization, which promotes the use of a particular eye.
  • Clear acrylic construction
Zebrafish bite test evaluates cerebral lateralization which promotes the use of a particular eye, according to the visual stimulus that is being fixated. Additionally, the bite test apparatus is employed to assess and correlate the biting habituation in Zebrafish to the preferential eye use, on viewing a novel object.
The apparatus can be conveniently attached to the inside of a home tank. The bite test apparatus stays connected to the main part of the tank with the help of a small opening that allows the fish to enter or leave the box during trials expediently.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty