Would you like to log in again?
Zebrafish Larvae Y-Maze
Supplier: Maze Engineers
4815
76670-120EA 1453.34 USD InStock
76670-120
Zebrafish Larvae Y-Maze
Animal Supplies Animal Environmental Enrichment
The Zebrafish larvae Y-Maze takes advantage of unique backlighting to allow for fine behavior task analysis in larvae.
  • Clear acrylic construction
  • Includes lid, chamber, and backlight
Similar to the Drosophila mazes, this apparatus comes with a start lane, bidirectional swimming pools, and a unique backlight for easy video tracking. An easy-to-use cover seals the pools and watertight chambers ensure a long service life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty